The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160302.1 | NP_001153774.1 | 14 - 15 | 1844 | 134 |
NM_001160306.1 | NP_001153778.1 | 13 - 14 | 1829 | 134 | |
NM_003895.3 | NP_003886.3 | 14 - 15 | 1836 | 134 | |
NM_203446.2 | NP_982271.2 | 14 - 15 | 1836 | 134 | |
XM_017028494.1 | XP_016883983.1 | 14 - 15 | 1826 | 134 | |
XM_017028495.1 | XP_016883984.1 | 13 - 14 | 1836 | 134 | |
XM_017028496.1 | XP_016883985.1 | 14 - 15 | 1826 | 134 | |
XM_017028497.1 | XP_016883986.1 | 14 - 15 | 1924 | 134 | |
XM_017028498.1 | XP_016883987.1 | 14 - 15 | 1826 | 134 | |
XM_017028499.1 | XP_016883988.1 | 13 - 14 | 1836 | 134 | |
XM_017028500.1 | XP_016883989.1 | 14 - 15 | 1826 | 134 | |
XM_017028501.1 | XP_016883990.1 | 14 - 15 | 1826 | 134 | |
XM_017028502.1 | XP_016883991.1 | 14 - 15 | 1826 | 134 | |
XM_017028503.1 | XP_016883992.1 | 14 - 15 | 1826 | 134 | |
XM_017028504.1 | XP_016883993.1 | 14 - 15 | 1826 | 134 | |
XM_017028505.1 | XP_016883994.1 | 13 - 14 | 1836 | 134 | |
GenBank mRNA | AB020717.2 | - | 14 - 15 | 1828 | 134 |
AF009039.1 | - | 14 - 15 | 1762 | 134 | |
AF009040.1 | - | 14 - 15 | 1762 | 134 | |
AK307762.1 | - | 14 - 15 | 1826 | 134 | |
BC019630.1 | - | 13 - 14 | 1811 | 134 | |
BC098395.1 | - | 13 - 14 | 1790 | 134 | |
BC136603.1 | - | 12 - 13 | 1725 | 134 |