Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNJ1
Entrez Gene ID
8867
Gene Name
synaptojanin 1
Gene Aliases
INPP5G, PARK20
Chromosome Location
Chr.21: 32628759 - 32728128 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160302.1 NP_001153774.1 22 - 23 2995 141
NM_001160306.1 NP_001153778.1 21 - 22 2980 141
NM_003895.3 NP_003886.3 22 - 23 2987 141
NM_203446.2 NP_982271.2 22 - 23 2987 141
XM_017028494.1 XP_016883983.1 22 - 23 2977 141
XM_017028495.1 XP_016883984.1 21 - 22 2987 141
XM_017028496.1 XP_016883985.1 22 - 23 2977 141
XM_017028497.1 XP_016883986.1 22 - 23 3075 141
XM_017028498.1 XP_016883987.1 22 - 23 2977 141
XM_017028499.1 XP_016883988.1 21 - 22 2987 141
XM_017028500.1 XP_016883989.1 22 - 23 2977 141
XM_017028501.1 XP_016883990.1 22 - 23 2977 141
XM_017028502.1 XP_016883991.1 22 - 23 2977 141
XM_017028503.1 XP_016883992.1 22 - 23 2977 141
XM_017028504.1 XP_016883993.1 22 - 23 2977 141
XM_017028505.1 XP_016883994.1 21 - 22 2987 141
GenBank mRNA AB020717.2 - 22 - 23 2979 141
AF009039.1 - 22 - 23 2913 141
AF009040.1 - 22 - 23 2913 141
AK307762.1 - 22 - 23 2977 141
BC098395.1 - 21 - 22 2941 141
BC136603.1 - 20 - 21 2876 141

View Full Product Details