Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB40
Entrez Gene ID
9923
Gene Name
zinc finger and BTB domain containing 40
Gene Aliases
ZNF923
Chromosome Location
Chr.1: 22428838 - 22531157 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083621.1 NP_001077090.1 15 - 16 3563 67
NM_014870.3 NP_055685.3 14 - 15 3275 67
XM_011542499.2 XP_011540801.1 14 - 15 3298 67
XM_011542500.2 XP_011540802.1 7 - 8 2548 67
XM_017003003.1 XP_016858492.1 12 - 13 2939 67
GenBank mRNA AB007947.2 - 14 - 15 3269 67
AK314747.1 - 4 - 5 977 67
BC114607.1 - 12 - 13 2811 67

View Full Product Details