The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083621.1 | NP_001077090.1 | 17 - 18 | 3811 | 61 |
NM_014870.3 | NP_055685.3 | 16 - 17 | 3523 | 61 | |
XM_011542499.2 | XP_011540801.1 | 16 - 17 | 3546 | 61 | |
XM_011542500.2 | XP_011540802.1 | 9 - 10 | 2796 | 61 | |
XM_017003003.1 | XP_016858492.1 | 14 - 15 | 3187 | 61 | |
GenBank mRNA | AB007947.2 | - | 16 - 17 | 3517 | 61 |
AK314747.1 | - | 6 - 7 | 1225 | 61 | |
BC114607.1 | - | 14 - 15 | 3059 | 61 |