The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114108.1 | NP_001107580.1 | 5 - 6 | 1122 | 59 |
XM_017001582.1 | XP_016857071.1 | 5 - 6 | 2256 | 59 | |
GenBank mRNA | AI923972.1 | - | 2 - 3 | 59 | |
AL049431.1 | - | 1 - 2 | 2011 | 59 |