The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033045.3 | NP_149034.2 | 4 - 5 | 961 | 80 |
XM_011538335.2 | XP_011536637.1 | 5 - 6 | 1076 | 80 | |
GenBank mRNA | AJ628420.1 | - | 4 - 5 | 907 | 80 |
AK314030.1 | - | 4 - 5 | 980 | 80 | |
BC069647.1 | - | 4 - 5 | 961 | 80 |