The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000251.2 | NP_000242.1 | 11 - 12 | 1882 | 75 |
NM_001258281.1 | NP_001245210.1 | 12 - 13 | 1698 | 75 | |
XM_005264332.3 | XP_005264389.2 | 11 - 12 | 1829 | 75 | |
XM_011532867.1 | XP_011531169.1 | 11 - 12 | 1829 | 75 | |
GenBank mRNA | AK222860.1 | - | 11 - 12 | 1819 | 75 |
AK223284.1 | - | 11 - 12 | 1792 | 75 | |
AK296831.1 | - | 10 - 11 | 1357 | 75 | |
AK297763.1 | - | 11 - 12 | 1585 | 75 | |
AK299667.1 | - | 10 - 11 | 1217 | 75 | |
AK304496.1 | - | 12 - 13 | 1698 | 75 | |
BC021566.1 | - | 11 - 12 | 1771 | 75 | |
BX649122.1 | - | 11 - 12 | 1777 | 75 | |
DQ648897.1 | - | 3 - 4 | 285 | 75 | |
L47574.1 | - | 11 - 12 | 1797 | 75 | |
L47575.1 | - | 11 - 12 | 1795 | 75 | |
L47576.1 | - | 11 - 12 | 1796 | 75 | |
L47577.1 | - | 11 - 12 | 1797 | 75 | |
L47578.1 | - | 11 - 12 | 1798 | 75 | |
L47579.1 | - | 11 - 12 | 1798 | 75 | |
L47580.1 | - | 11 - 12 | 1797 | 75 | |
L47581.1 | - | 11 - 12 | 1797 | 75 | |
L47582.1 | - | 11 - 12 | 1797 | 75 | |
U03911.1 | - | 11 - 12 | 1760 | 75 | |
U04045.1 | - | 11 - 12 | 1825 | 75 |