Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSIG10
Entrez Gene ID
54621
Gene Name
V-set and immunoglobulin domain containing 10
Gene Aliases
-
Chromosome Location
Chr.12: 118063593 - 118104402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019086.5 NP_061959.2 2 - 3 694 67
XM_011538502.1 XP_011536804.1 2 - 3 694 67
XM_017019504.1 XP_016874993.1 3 - 4 733 67
XM_017019505.1 XP_016874994.1 2 - 3 339 67
XM_017019506.1 XP_016874995.1 2 - 3 828 67
GenBank mRNA AK000681.1 - 2 - 3 724 67
BC028698.2 - 2 - 3 638 67
BC034471.2 - 2 - 3 633 67

View Full Product Details