The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003730.4 | NP_003721.2 | 4 - 5 | 669 | 75 |
XM_017011397.1 | XP_016866886.1 | 5 - 6 | 448 | 75 | |
XM_017011398.1 | XP_016866887.1 | 3 - 4 | 570 | 75 | |
XM_017011399.1 | XP_016866888.1 | 4 - 5 | 630 | 75 | |
GenBank mRNA | AJ419865.1 | - | 4 - 5 | 623 | 75 |
AK315467.1 | - | 4 - 5 | 630 | 75 | |
BC001660.2 | - | 4 - 5 | 595 | 75 | |
BC001819.2 | - | 4 - 5 | 595 | 75 | |
BC036282.1 | - | 4 - 5 | 656 | 75 | |
BC039713.1 | - | 4 - 5 | 598 | 75 | |
BC051912.1 | - | 4 - 5 | 608 | 75 | |
U85625.2 | - | 4 - 5 | 626 | 75 |