The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_004859.1 | 2 - 3 | 1401 | 83 | |
GenBank mRNA | AA280726.1 | - | 1 - 2 | 105 | 83 |
AI218957.1 | - | 1 - 2 | 105 | 83 |