The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001112.3 | NP_001103.1 | 9 - 10 | 2189 | 53 |
NM_001160230.1 | NP_001153702.1 | 9 - 10 | 2189 | 53 | |
NM_015833.3 | NP_056648.1 | 10 - 11 | 2309 | 53 | |
NM_015834.3 | NP_056649.1 | 10 - 11 | 2309 | 53 | |
NR_027672.1 | 8 - 9 | 1254 | 53 | ||
NR_027673.1 | 10 - 11 | 2307 | 53 | ||
NR_027674.1 | 9 - 10 | 2189 | 53 | ||
NR_073200.1 | 10 - 11 | 2309 | 53 | ||
XM_006723954.2 | XP_006724017.1 | 11 - 12 | 2381 | 53 | |
XM_011529425.2 | XP_011527727.1 | 11 - 12 | 3501 | 53 | |
XM_011529432.1 | XP_011527734.1 | 9 - 10 | 1222 | 53 | |
XM_017028242.1 | XP_016883731.1 | 9 - 10 | 3977 | 53 | |
XM_017028243.1 | XP_016883732.1 | 12 - 13 | 7466 | 53 | |
XM_017028244.1 | XP_016883733.1 | 9 - 10 | 3975 | 53 | |
XM_017028245.1 | XP_016883734.1 | 8 - 9 | 3810 | 53 | |
XM_017028246.1 | XP_016883735.1 | 12 - 13 | 7466 | 53 | |
XM_017028247.1 | XP_016883736.1 | 11 - 12 | 7348 | 53 | |
XM_017028248.1 | XP_016883737.1 | 12 - 13 | 2290 | 53 | |
XM_017028249.1 | XP_016883738.1 | 13 - 14 | 7509 | 53 | |
XM_017028250.1 | XP_016883739.1 | 12 - 13 | 7419 | 53 | |
XM_017028251.1 | XP_016883740.1 | 9 - 10 | 2091 | 53 | |
XM_017028252.1 | XP_016883741.1 | 9 - 10 | 3977 | 53 | |
XM_017028253.1 | XP_016883742.1 | 8 - 9 | 3805 | 53 | |
XM_017028254.1 | XP_016883743.1 | 11 - 12 | 7348 | 53 | |
XM_017028255.1 | XP_016883744.1 | 10 - 11 | 7226 | 53 | |
XM_017028256.1 | XP_016883745.1 | 9 - 10 | 2021 | 53 | |
GenBank mRNA | AB194369.1 | - | 8 - 9 | 1232 | 53 |
AB194370.1 | - | 9 - 10 | 2167 | 53 | |
AB194371.1 | - | 9 - 10 | 2167 | 53 | |
AB194372.2 | - | 9 - 10 | 2167 | 53 | |
AB194373.1 | - | 9 - 10 | 2167 | 53 | |
AF001042.1 | - | 8 - 9 | 1889 | 53 | |
AF525422.1 | - | 10 - 11 | 2285 | 53 | |
AF533142.1 | - | 9 - 10 | 2167 | 53 | |
AY135659.1 | - | 10 - 11 | 2285 | 53 | |
BC065545.1 | - | 9 - 10 | 2026 | 53 | |
U76420.1 | - | 9 - 10 | 2167 | 53 | |
U76421.1 | - | 10 - 11 | 2287 | 53 | |
U76422.1 | - | 10 - 11 | 2287 | 53 | |
U82120.1 | - | 9 - 10 | 1994 | 53 | |
U82121.1 | - | 8 - 9 | 1874 | 53 | |
X99227.1 | - | 9 - 10 | 2021 | 53 | |
X99383.1 | - | 8 - 9 | 1901 | 53 |