Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAGLN2
Entrez Gene ID
8407
Gene Name
transgelin 2
Gene Aliases
HA1756
Chromosome Location
Chr.1: 159918107 - 159925542 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277224.1 NP_001264153.1 1 - 2 279 73
GenBank mRNA AY007127.1 - 1 - 2 279 73

View Full Product Details