Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFWD3
Entrez Gene ID
55159
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
RNF201
Chromosome Location
Chr.16: 74621396 - 74666892 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018124.3 NP_060594.3 7 - 8 1289 100
XM_005256021.4 XP_005256078.1 8 - 9 1422 100
XM_005256022.4 XP_005256079.1 7 - 8 2172 100
XM_006721228.3 XP_006721291.1 8 - 9 1422 100
XM_011523191.2 XP_011521493.1 7 - 8 1242 100
XM_017023391.1 XP_016878880.1 8 - 9 1338 100
XM_017023392.1 XP_016878881.1 7 - 8 1286 100
GenBank mRNA AK001382.1 - 2 - 3 191 100
AK291200.1 - 8 - 9 1389 100
AK315786.1 - 4 - 5 433 100
BC002574.2 - 5 - 6 666 100
BC059371.1 - 7 - 8 1238 100

View Full Product Details