The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020435.3 | NP_065168.2 | 1 - 2 | 293 | 75 |
GenBank mRNA | BC035840.1 | - | 1 - 2 | 47 | 75 |
BC089439.1 | - | 1 - 2 | 293 | 75 | |
CF132637.1 | - | 1 - 2 | 75 | ||
EU433401.1 | - | 1 - 2 | 77 | 75 |