The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001388.4 | NP_001379.1 | 7 - 8 | 785 | 73 |
XM_005256499.2 | XP_005256556.1 | 7 - 8 | 767 | 73 | |
XM_005256500.2 | XP_005256557.1 | 7 - 8 | 762 | 73 | |
XM_011523704.1 | XP_011522006.1 | 7 - 8 | 767 | 73 | |
XM_011523705.1 | XP_011522007.1 | 7 - 8 | 767 | 73 | |
XM_011523706.1 | XP_011522008.1 | 7 - 8 | 696 | 73 | |
XM_011523707.1 | XP_011522009.1 | 7 - 8 | 742 | 73 | |
XM_017024296.1 | XP_016879785.1 | 7 - 8 | 696 | 73 | |
XM_017024297.1 | XP_016879786.1 | 7 - 8 | 742 | 73 | |
GenBank mRNA | AB209340.1 | - | 3 - 4 | 4146 | 73 |
AK297569.1 | - | 6 - 7 | 1314 | 73 | |
AK313362.1 | - | 7 - 8 | 704 | 73 | |
BC000493.2 | - | 7 - 8 | 683 | 73 | |
BT006976.1 | - | 7 - 8 | 628 | 73 | |
X80754.1 | - | 7 - 8 | 675 | 73 |