Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PALB2
Entrez Gene ID
79728
Gene Name
partner and localizer of BRCA2
Gene Aliases
FANCN, PNCA3
Chromosome Location
Chr.16: 23603162 - 23641357 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024675.3 NP_078951.2 7 - 8 2948 79
XM_011545946.2 XP_011544248.1 7 - 8 3753 79
XM_011545947.2 XP_011544249.1 7 - 8 3718 79
XM_011545948.2 XP_011544250.1 8 - 9 3040 79
XM_017023671.1 XP_016879160.1 7 - 8 3718 79
XM_017023672.1 XP_016879161.1 7 - 8 2948 79
XM_017023673.1 XP_016879162.1 7 - 8 2948 79
GenBank mRNA AK025469.1 - 4 - 5 1325 79
AK299152.1 - 7 - 8 2845 79
AL834425.1 - 4 - 5 2231 79
BC044254.2 - 7 - 8 2875 79
BX647125.1 - 7 - 8 2859 79
BX647130.1 - 7 - 8 2901 79
CR749637.1 - 7 - 8 2899 79

View Full Product Details