The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242338.2 | NP_001229267.2 | 21 - 22 | 3303 | 140 |
NR_125717.1 | 22 - 23 | 3387 | 140 | ||
XM_011532919.2 | XP_011531221.1 | 22 - 23 | 3345 | 140 | |
XM_011532921.2 | XP_011531223.1 | 22 - 23 | 3345 | 140 | |
XM_017004375.1 | XP_016859864.1 | 21 - 22 | 3303 | 140 | |
GenBank mRNA | AB033066.1 | - | 5 - 6 | 801 | 140 |
BC171846.1 | - | 22 - 23 | 3334 | 140 |