Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GREB1
Entrez Gene ID
9687
Gene Name
growth regulation by estrogen in breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.2: 11534116 - 11642788 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014668.3 NP_055483.2 23 - 24 4445 77
XM_005246192.4 XP_005246249.1 23 - 24 4467 77
XM_005246196.3 XP_005246253.1 7 - 8 1520 77
XM_011510418.2 XP_011508720.1 23 - 24 4303 77
XM_011510419.2 XP_011508721.1 22 - 23 4303 77
XM_011510422.2 XP_011508724.1 8 - 9 1615 77
GenBank mRNA AB011147.1 - 6 - 7 1248 77
BC054502.1 - 6 - 7 1257 77

View Full Product Details