The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003654.2 | 1 - 2 | 834 | 66 | |
GenBank mRNA | AK290489.1 | - | 1 - 2 | 844 | 66 |
BC011547.1 | - | 1 - 2 | 816 | 66 | |
BC012929.1 | - | 1 - 2 | 816 | 66 | |
BC063836.1 | - | 1 - 2 | 790 | 66 | |
BC093922.1 | - | 1 - 2 | 467 | 66 | |
BC093924.1 | - | 1 - 2 | 467 | 66 | |
BC143241.1 | - | 1 - 2 | 467 | 66 |