Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH3
Entrez Gene ID
55567
Gene Name
dynein axonemal heavy chain 3
Gene Aliases
DLP3, DNAHC3B, HEL-36, Hsadhc3, dnahc3-b
Chromosome Location
Chr.16: 20933111 - 21159537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017539.2 NP_060009.1 26 - 27 3816 71
XM_006721062.1 XP_006721125.1 26 - 27 3954 71
XM_011545882.2 XP_011544184.1 27 - 28 4394 71
XM_011545883.1 XP_011544185.1 27 - 28 4162 71
XM_011545885.2 XP_011544187.1 27 - 28 4394 71
XM_011545886.2 XP_011544188.1 19 - 20 2769 71
XM_011545888.2 XP_011544190.1 9 - 10 1213 71
XM_011545889.2 XP_011544191.1 9 - 10 1177 71
XM_017023426.1 XP_016878915.1 27 - 28 4394 71
XM_017023427.1 XP_016878916.1 25 - 26 4151 71
XM_017023428.1 XP_016878917.1 27 - 28 4394 71
XM_017023429.1 XP_016878918.1 27 - 28 4395 71
XM_017023431.1 XP_016878920.1 27 - 28 4395 71
XM_017023432.1 XP_016878921.1 26 - 27 3954 71
GenBank mRNA AF494040.1 - 26 - 27 3816 71
AK125907.1 - 3 - 4 533 71

View Full Product Details