The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 32 - 33 | 4633 | 64 |
XM_006721062.1 | XP_006721125.1 | 32 - 33 | 4771 | 64 | |
XM_011545882.2 | XP_011544184.1 | 33 - 34 | 5211 | 64 | |
XM_011545883.1 | XP_011544185.1 | 33 - 34 | 4979 | 64 | |
XM_011545885.2 | XP_011544187.1 | 33 - 34 | 5211 | 64 | |
XM_011545886.2 | XP_011544188.1 | 25 - 26 | 3586 | 64 | |
XM_011545888.2 | XP_011544190.1 | 15 - 16 | 2030 | 64 | |
XM_011545889.2 | XP_011544191.1 | 15 - 16 | 1994 | 64 | |
XM_017023426.1 | XP_016878915.1 | 33 - 34 | 5211 | 64 | |
XM_017023427.1 | XP_016878916.1 | 31 - 32 | 4968 | 64 | |
XM_017023428.1 | XP_016878917.1 | 33 - 34 | 5211 | 64 | |
XM_017023429.1 | XP_016878918.1 | 33 - 34 | 5212 | 64 | |
XM_017023431.1 | XP_016878920.1 | 33 - 34 | 5212 | 64 | |
XM_017023432.1 | XP_016878921.1 | 32 - 33 | 4771 | 64 | |
GenBank mRNA | AF494040.1 | - | 32 - 33 | 4633 | 64 |
AK125907.1 | - | 9 - 10 | 1350 | 64 | |
Z83805.1 | - | 3 - 4 | 349 | 64 |