The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017539.2 | NP_060009.1 | 59 - 60 | 11653 | 86 |
XM_006721062.1 | XP_006721125.1 | 59 - 60 | 11791 | 86 | |
XM_011545882.2 | XP_011544184.1 | 60 - 61 | 12231 | 86 | |
XM_011545883.1 | XP_011544185.1 | 60 - 61 | 11999 | 86 | |
XM_011545886.2 | XP_011544188.1 | 52 - 53 | 10606 | 86 | |
XM_011545888.2 | XP_011544190.1 | 42 - 43 | 9050 | 86 | |
XM_011545889.2 | XP_011544191.1 | 42 - 43 | 9014 | 86 | |
XM_017023426.1 | XP_016878915.1 | 59 - 60 | 12096 | 86 | |
XM_017023427.1 | XP_016878916.1 | 58 - 59 | 11988 | 86 | |
XM_017023428.1 | XP_016878917.1 | 57 - 58 | 11652 | 86 | |
XM_017023430.1 | XP_016878919.1 | 18 - 19 | 5673 | 86 | |
XM_017023432.1 | XP_016878921.1 | 59 - 60 | 11791 | 86 | |
GenBank mRNA | AF494040.1 | - | 59 - 60 | 11653 | 86 |
AK125952.1 | - | 12 - 13 | 4006 | 86 | |
AL096732.1 | - | 7 - 8 | 2200 | 86 |