Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC7A2
Entrez Gene ID
6542
Gene Name
solute carrier family 7 member 2
Gene Aliases
ATRC2, CAT2, HCAT2
Chromosome Location
Chr.8: 17494349 - 17570568 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AA724796.1 - 2 - 3 64 63

View Full Product Details