Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location
Chr.9: 110873252 - 111039259 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_057159.2 NP_476500.1 2 - 3 325 67
XM_005251781.3 XP_005251838.1 2 - 3 470 67
XM_005251782.3 XP_005251839.1 2 - 3 564 67
XM_017014383.1 XP_016869872.1 2 - 3 364 67
XM_017014384.1 XP_016869873.1 3 - 4 738 67
XM_017014385.1 XP_016869874.1 4 - 5 331 67
XM_017014386.1 XP_016869875.1 4 - 5 631 67
XM_017014387.1 XP_016869876.1 4 - 5 367 67
XM_017014388.1 XP_016869877.1 4 - 5 550 67
XM_017014389.1 XP_016869878.1 3 - 4 293 67
XM_017014390.1 XP_016869879.1 4 - 5 328 67
XM_017014391.1 XP_016869880.1 4 - 5 352 67
XM_017014392.1 XP_016869881.1 3 - 4 595 67
XM_017014393.1 XP_016869882.1 4 - 5 391 67
XM_017014394.1 XP_016869883.1 3 - 4 520 67
XM_017014395.1 XP_016869884.1 3 - 4 348 67
XM_017014396.1 XP_016869885.1 2 - 3 290 67
XM_017014397.1 XP_016869886.1 3 - 4 221 67
XM_017014398.1 XP_016869887.1 3 - 4 464 67
XM_017014399.1 XP_016869888.1 3 - 4 219 67
XM_017014400.1 XP_016869889.1 4 - 5 580 67
XM_017014401.1 XP_016869890.1 3 - 4 251 67
XM_017014402.1 XP_016869891.1 3 - 4 308 67
XM_017014403.1 XP_016869892.1 4 - 5 306 67
XM_017014404.1 XP_016869893.1 2 - 3 504 67
XM_017014405.1 XP_016869894.1 4 - 5 493 67
XM_017014406.1 XP_016869895.1 3 - 4 227 67
XM_017014407.1 XP_016869896.1 3 - 4 283 67
GenBank mRNA AK290287.1 - 3 - 4 600 67
BC030615.2 - 2 - 3 154 67
BC036034.1 - 2 - 3 284 67
BC073167.1 - 2 - 3 166 67
BI461641.1 - 2 - 3 292 67

View Full Product Details