Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYSL2
Entrez Gene ID
1808
Gene Name
dihydropyrimidinase like 2
Gene Aliases
CRMP-2, CRMP2, DHPRP2, DRP-2, DRP2, N2A3, ULIP-2, ULIP2
Chromosome Location
Chr.8: 26514193 - 26658177 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197293.2 NP_001184222.1 13 - 14 2078 118
NM_001244604.1 NP_001231533.1 13 - 14 1720 118
NM_001386.5 NP_001377.1 13 - 14 2061 118
GenBank mRNA AB209195.1 - 13 - 14 1799 118
AK092377.1 - 5 - 6 811 118
AK223557.1 - 13 - 14 1924 118
AK291287.1 - 13 - 14 1842 118
AK299077.1 - 13 - 14 1713 118
BC056408.1 - 13 - 14 1915 118
BC067109.1 - 13 - 14 1945 118
BX647115.1 - 13 - 14 1989 118
D78013.1 - 13 - 14 1904 118
U17279.1 - 13 - 14 1701 118

View Full Product Details