The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013309.5 | NP_037441.2 | 6 - 7 | 1314 | 163 |
XM_011521997.2 | XP_011520299.1 | 6 - 7 | 1818 | 163 | |
XM_017022560.1 | XP_016878049.1 | 5 - 6 | 1120 | 163 | |
GenBank mRNA | AF025409.1 | - | 5 - 6 | 1002 | 163 |
AK290874.1 | - | 5 - 6 | 1120 | 163 | |
AY029482.1 | - | 2 - 3 | 279 | 163 | |
BC026089.2 | - | 6 - 7 | 1314 | 163 |