The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002688.5 | NP_002679.2 | 1 - 2 | 171 | 63 |
GenBank mRNA | AK295940.1 | - | 1 - 2 | 40 | 63 |
BC025261.1 | - | 1 - 2 | 135 | 63 | |
CR456545.1 | - | 1 - 2 | 51 | 63 | |
KJ534756.1 | - | 1 - 2 | 46 | 63 | |
KJ534757.1 | - | 1 - 2 | 46 | 63 | |
U74628.1 | - | 1 - 2 | 133 | 63 | |
U85431.1 | - | 1 - 2 | 112 | 63 | |
Y11593.1 | - | 1 - 2 | 91 | 63 |