The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160184.1 | NP_001153656.1 | 12 - 13 | 1363 | 114 |
NM_032129.2 | NP_115505.2 | 13 - 14 | 1468 | 114 | |
XM_006710944.3 | XP_006711007.2 | 13 - 14 | 2181 | 114 | |
XM_011542248.2 | XP_011540550.2 | 12 - 13 | 2301 | 114 | |
XM_017002474.1 | XP_016857963.1 | 11 - 12 | 2160 | 114 | |
GenBank mRNA | AL136730.1 | - | 13 - 14 | 1468 | 114 |
BC101386.2 | - | 12 - 13 | 114 | ||
BC101387.2 | - | 12 - 13 | 1619 | 114 |