Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AL109783.1 - 1 1267 Chr. 21 - 32259795 - 32261585 [-] on Build GRCh38 64


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 159 LINC00159 C21orf44, NCRNA00159 -173.5kb Sense Chr. - 32080481 - 32086295 on Build GRCh38
Downstream MIS18 kinetochore protein A MIS18A B28, C21orf45, C21orf46, FASP1, MIS18alpha, hMis18alpha 6.6kb Sense Chr. - 32244759 - 32283954 on Build GRCh38

View Full Product Details