The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017864.3 | NP_060334.2 | 23 - 24 | 145 | |
NR_073444.1 | 25 - 26 | 145 | |||
NR_073445.1 | 24 - 25 | 145 | |||
XM_017013616.1 | XP_016869105.1 | 23 - 24 | 145 | ||
XM_017013617.1 | XP_016869106.1 | 21 - 22 | 145 | ||
XM_017013618.1 | XP_016869107.1 | 20 - 21 | 145 | ||
XM_017013619.1 | XP_016869108.1 | 23 - 24 | 145 | ||
GenBank mRNA | AB161944.1 | - | 23 - 24 | 2813 | 145 |
AK000537.1 | - | 13 - 14 | 145 | ||
AK001403.1 | - | 16 - 17 | 1769 | 145 | |
AK091278.1 | - | 22 - 23 | 2566 | 145 | |
AK225533.1 | - | 13 - 14 | 145 | ||
BC050536.1 | - | 15 - 16 | 1617 | 145 | |
BC064915.1 | - | 15 - 16 | 145 | ||
BC136754.1 | - | 23 - 24 | 145 | ||
BC144396.1 | - | 25 - 26 | 145 | ||
BC144397.1 | - | 24 - 25 | 145 | ||
BM474137.1 | - | 3 - 4 | 145 |