The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017864.3 | NP_060334.2 | 25 - 26 | 2963 | 153 |
NR_073444.1 | 27 - 28 | 3137 | 153 | ||
NR_073445.1 | 26 - 27 | 3054 | 153 | ||
XM_017013617.1 | XP_016869106.1 | 23 - 24 | 2744 | 153 | |
XM_017013618.1 | XP_016869107.1 | 22 - 23 | 2516 | 153 | |
XM_017013619.1 | XP_016869108.1 | 25 - 26 | 2922 | 153 | |
GenBank mRNA | AB161944.1 | - | 25 - 26 | 2947 | 153 |
AK000537.1 | - | 15 - 16 | 1517 | 153 | |
AK001403.1 | - | 18 - 19 | 1903 | 153 | |
AK091278.1 | - | 24 - 25 | 2700 | 153 | |
AK225533.1 | - | 15 - 16 | 1517 | 153 | |
BC050536.1 | - | 17 - 18 | 1751 | 153 | |
BC064915.1 | - | 17 - 18 | 1792 | 153 | |
BC136754.1 | - | 25 - 26 | 2877 | 153 | |
BC144396.1 | - | 27 - 28 | 3051 | 153 | |
BC144397.1 | - | 26 - 27 | 2968 | 153 | |
BM474137.1 | - | 5 - 6 | 398 | 153 | |
BX538203.1 | - | 20 - 21 | 2235 | 153 |