The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017864.3 | NP_060334.2 | 3 - 4 | 587 | 82 |
NR_073444.1 | 3 - 4 | 587 | 82 | ||
NR_073445.1 | 3 - 4 | 587 | 82 | ||
XM_017013616.1 | XP_016869105.1 | 3 - 4 | 587 | 82 | |
XM_017013617.1 | XP_016869106.1 | 3 - 4 | 587 | 82 | |
XM_017013618.1 | XP_016869107.1 | 2 - 3 | 404 | 82 | |
XM_017013619.1 | XP_016869108.1 | 3 - 4 | 577 | 82 | |
GenBank mRNA | AB161944.1 | - | 3 - 4 | 571 | 82 |
AK091278.1 | - | 2 - 3 | 324 | 82 | |
BC136754.1 | - | 3 - 4 | 501 | 82 | |
BC144396.1 | - | 3 - 4 | 501 | 82 | |
BC144397.1 | - | 3 - 4 | 501 | 82 |