Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
Chromosome Location
Chr.8: 94823290 - 94894664 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017864.3 NP_060334.2 4 - 5 653 140
NR_073444.1 4 - 5 653 140
NR_073445.1 4 - 5 653 140
XM_017013616.1 XP_016869105.1 4 - 5 653 140
XM_017013617.1 XP_016869106.1 4 - 5 653 140
XM_017013618.1 XP_016869107.1 3 - 4 470 140
XM_017013619.1 XP_016869108.1 4 - 5 643 140
GenBank mRNA AB161944.1 - 4 - 5 637 140
AK091278.1 - 3 - 4 390 140
BC136754.1 - 4 - 5 567 140
BC144396.1 - 4 - 5 567 140
BC144397.1 - 4 - 5 567 140

View Full Product Details