Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM91A1
Entrez Gene ID
157769
Gene Name
family with sequence similarity 91 member A1
Gene Aliases
-
Chromosome Location
Chr.8: 123768439 - 123815452 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317917.1 NP_001304846.1 14 - 15 1552 159
NM_001317918.1 NP_001304847.1 14 - 15 1545 159
NM_144963.3 NP_659400.2 14 - 15 1545 159
GenBank mRNA AK074370.1 - 9 - 10 852 159
AK300603.1 - 14 - 15 1528 159
AL832830.1 - 3 - 4 276 159
BC144630.1 - 14 - 15 1345 159
DQ228141.1 - 14 - 15 1342 159

View Full Product Details