The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015204.2 | NP_056019.1 | 2 - 3 | 1273 | 129 |
XM_006715659.1 | XP_006715722.1 | 2 - 3 | 1272 | 129 | |
XM_006715660.1 | XP_006715723.1 | 2 - 3 | 1272 | 129 | |
XM_006715662.1 | XP_006715725.1 | 2 - 3 | 1272 | 129 | |
XM_011515193.2 | XP_011513495.1 | 2 - 3 | 3597 | 129 | |
XM_011515194.1 | XP_011513496.1 | 2 - 3 | 1038 | 129 | |
GenBank mRNA | AB023177.2 | - | 1 - 2 | 557 | 129 |
AK092252.1 | - | 2 - 3 | 1273 | 129 |