Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THSD7A
Entrez Gene ID
221981
Gene Name
thrombospondin type 1 domain containing 7A
Gene Aliases
-
Chromosome Location
Chr.7: 11370435 - 11832198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015204.2 NP_056019.1 4 - 5 1708 112
XM_006715659.1 XP_006715722.1 4 - 5 1707 112
XM_006715660.1 XP_006715723.1 4 - 5 1707 112
XM_006715662.1 XP_006715725.1 4 - 5 1707 112
XM_011515193.2 XP_011513495.1 4 - 5 4032 112
XM_011515194.1 XP_011513496.1 4 - 5 1473 112
GenBank mRNA AB023177.2 - 3 - 4 992 112
AK092252.1 - 4 - 5 1708 112

View Full Product Details