The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003718.4 | NP_003709.3 | 6 - 7 | 2828 | 128 |
NM_031267.3 | NP_112557.2 | 6 - 7 | 2828 | 128 | |
XM_011515597.2 | XP_011513899.1 | 6 - 7 | 3687 | 128 | |
XM_017012750.1 | XP_016868239.1 | 6 - 7 | 3687 | 128 | |
XM_017012751.1 | XP_016868240.1 | 6 - 7 | 3687 | 128 | |
GenBank mRNA | AJ297709.1 | - | 6 - 7 | 2678 | 128 |
AJ297710.1 | - | 6 - 7 | 2678 | 128 | |
BC001274.1 | - | 5 - 6 | 1098 | 128 | |
M80629.1 | - | 6 - 7 | 1475 | 128 |