The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042599.1 | NP_001036064.1 | 13 - 14 | 1725 | 89 |
NM_005235.2 | NP_005226.1 | 13 - 14 | 1725 | 89 | |
XM_005246376.2 | XP_005246433.1 | 13 - 14 | 2244 | 89 | |
XM_005246377.2 | XP_005246434.1 | 13 - 14 | 2244 | 89 | |
XM_006712364.2 | XP_006712427.1 | 13 - 14 | 2244 | 89 | |
XM_017003577.1 | XP_016859066.1 | 14 - 15 | 2322 | 89 | |
XM_017003578.1 | XP_016859067.1 | 14 - 15 | 2322 | 89 | |
XM_017003579.1 | XP_016859068.1 | 14 - 15 | 2322 | 89 | |
XM_017003580.1 | XP_016859069.1 | 14 - 15 | 2322 | 89 | |
XM_017003581.1 | XP_016859070.1 | 14 - 15 | 2322 | 89 | |
XM_017003582.1 | XP_016859071.1 | 9 - 10 | 1357 | 89 | |
GenBank mRNA | AB209697.1 | - | 3 - 4 | 429 | 89 |
AK308606.1 | - | 13 - 14 | 1681 | 89 | |
BC112199.1 | - | 13 - 14 | 1666 | 89 | |
BC143741.1 | - | 13 - 14 | 1654 | 89 | |
BC143747.1 | - | 13 - 14 | 1654 | 89 | |
BC143749.1 | - | 13 - 14 | 1654 | 89 | |
KT310076.1 | - | 2 - 3 | 412 | 89 | |
L07868.1 | - | 13 - 14 | 1660 | 89 |