The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005235.2 | NP_005226.1 | 26 - 27 | 3282 | 70 |
XM_005246376.2 | XP_005246433.1 | 26 - 27 | 3771 | 70 | |
XM_006712364.2 | XP_006712427.1 | 27 - 28 | 3846 | 70 | |
XM_017003577.1 | XP_016859066.1 | 28 - 29 | 3924 | 70 | |
XM_017003578.1 | XP_016859067.1 | 27 - 28 | 3879 | 70 | |
XM_017003580.1 | XP_016859069.1 | 27 - 28 | 3849 | 70 | |
XM_017003582.1 | XP_016859071.1 | 23 - 24 | 2959 | 70 | |
GenBank mRNA | BC112199.1 | - | 26 - 27 | 3223 | 70 |
BC143747.1 | - | 26 - 27 | 3181 | 70 | |
L07868.1 | - | 26 - 27 | 3217 | 70 |