Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
Chromosome Location
Chr.12: 107317168 - 107659642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017523.1 NP_001017523.1 9 - 10 1289 84
NM_001018072.1 NP_001018082.1 11 - 12 3149 84
XM_005268645.3 XP_005268702.1 11 - 12 1517 84
XM_011537909.2 XP_011536211.1 11 - 12 2072 84
XM_011537910.2 XP_011536212.1 11 - 12 2026 84
XM_017018807.1 XP_016874296.1 10 - 11 4299 84
GenBank mRNA AK122818.1 - 11 - 12 1586 84
AK124835.1 - 11 - 12 1617 84
AK127295.1 - 9 - 10 1289 84
AK128445.1 - 3 - 4 528 84
AY373588.1 - 7 - 8 983 84
BC146824.1 - 9 - 10 2791 84

View Full Product Details