The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001017523.1 | NP_001017523.1 | 12 - 13 | 1585 | 80 |
NM_001018072.1 | NP_001018082.1 | 14 - 15 | 3445 | 80 | |
XM_005268645.3 | XP_005268702.1 | 14 - 15 | 1813 | 80 | |
XM_011537909.2 | XP_011536211.1 | 14 - 15 | 2368 | 80 | |
XM_011537910.2 | XP_011536212.1 | 14 - 15 | 2322 | 80 | |
XM_017018807.1 | XP_016874296.1 | 13 - 14 | 4595 | 80 | |
GenBank mRNA | AK091276.1 | - | 4 - 5 | 420 | 80 |
AK122818.1 | - | 14 - 15 | 1882 | 80 | |
AK124835.1 | - | 14 - 15 | 1913 | 80 | |
AK127012.1 | - | 11 - 12 | 1355 | 80 | |
AK127295.1 | - | 12 - 13 | 1585 | 80 | |
AK128445.1 | - | 6 - 7 | 824 | 80 | |
AY373588.1 | - | 10 - 11 | 1279 | 80 | |
BC093627.1 | - | 3 - 4 | 251 | 80 | |
BC093629.1 | - | 3 - 4 | 251 | 80 | |
BC101561.1 | - | 3 - 4 | 251 | 80 | |
BC101563.1 | - | 3 - 4 | 251 | 80 |