The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018072.1 | NP_001018082.1 | 1 - 2 | 1653 | 72 |
XM_017018807.1 | XP_016874296.1 | 1 - 2 | 2905 | 72 | |
GenBank mRNA | AK122818.1 | - | 1 - 2 | 152 | 72 |
AK124835.1 | - | 1 - 2 | 121 | 72 | |
BC146824.1 | - | 1 - 2 | 1652 | 72 |