Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
Chromosome Location
Chr.12: 107317168 - 107659642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017523.1 NP_001017523.1 6 - 7 758 87
NM_001018072.1 NP_001018082.1 8 - 9 2618 87
XM_005268645.3 XP_005268702.1 8 - 9 986 87
XM_011537909.2 XP_011536211.1 8 - 9 1541 87
XM_011537910.2 XP_011536212.1 8 - 9 1495 87
XM_017018807.1 XP_016874296.1 7 - 8 3768 87
GenBank mRNA AK122818.1 - 8 - 9 1055 87
AK124835.1 - 8 - 9 1086 87
AK127012.1 - 5 - 6 528 87
AK127295.1 - 6 - 7 758 87
AY373588.1 - 4 - 5 452 87

View Full Product Details