Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
Chromosome Location
Chr.12: 107317168 - 107659642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017523.1 NP_001017523.1 7 - 8 887 78
NM_001018072.1 NP_001018082.1 9 - 10 2747 78
XM_005268645.3 XP_005268702.1 9 - 10 1115 78
XM_011537909.2 XP_011536211.1 9 - 10 1670 78
XM_011537910.2 XP_011536212.1 9 - 10 1624 78
XM_017018807.1 XP_016874296.1 8 - 9 3897 78
GenBank mRNA AK122818.1 - 9 - 10 1184 78
AK124835.1 - 9 - 10 1215 78
AK127012.1 - 6 - 7 657 78
AK127295.1 - 7 - 8 887 78
AK128445.1 - 1 - 2 126 78
AY373588.1 - 5 - 6 581 78

View Full Product Details