The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017612.4 | NP_060082.2 | 3 - 4 | 547 | 158 |
XM_006719491.2 | XP_006719554.1 | 3 - 4 | 463 | 158 | |
GenBank mRNA | AK027702.1 | - | 3 - 4 | 416 | 158 |
AK074638.1 | - | 3 - 4 | 445 | 158 | |
AL157433.1 | - | 2 - 3 | 1841 | 158 | |
BC065918.1 | - | 3 - 4 | 386 | 158 |