Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WFDC6
Entrez Gene ID
140870
Gene Name
WAP four-disulfide core domain 6
Gene Aliases
C20orf171, HEL-S-295, WAP6, dJ461P17.11
Chromosome Location
Chr.20: 45534197 - 45539495 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080827.1 NP_543017.1 2 - 3 315 74
GenBank mRNA AF411861.1 - 2 - 3 315 74
BC101600.1 - 2 - 3 310 74
BC101604.1 - 2 - 3 310 74
GQ472217.1 - 2 - 3 310 74

View Full Product Details