The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175841.2 | NP_787035.1 | 3 - 4 | 662 | 64 |
GenBank mRNA | AY033891.1 | - | 3 - 4 | 506 | 64 |