Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS16
Entrez Gene ID
64601
Gene Name
VPS16, CORVET/HOPS core subunit
Gene Aliases
hVPS16
Chromosome Location
Chr.20: 2840697 - 2866732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022575.3 NP_072097.2 16 - 17 1683 114
NM_080413.2 NP_536338.1 12 - 13 1251 114
GenBank mRNA AF308801.1 - 16 - 17 1619 114
AK055787.1 - 12 - 13 1221 114
AL834401.1 - 12 - 13 1171 114
BC012422.1 - 4 - 5 334 114
BC021291.2 - 2 - 3 202 114
BC073959.1 - 4 - 5 753 114

View Full Product Details