The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002267.3 | NP_002258.2 | 10 - 11 | 1186 | 131 |
XM_017020561.1 | XP_016876050.1 | 10 - 11 | 869 | 131 | |
GenBank mRNA | AF005263.1 | - | 10 - 11 | 783 | 131 |
AK223480.1 | - | 10 - 11 | 808 | 131 | |
AK290528.1 | - | 10 - 11 | 846 | 131 | |
AK291000.1 | - | 10 - 11 | 810 | 131 | |
BC017355.2 | - | 10 - 11 | 795 | 131 | |
BC024202.2 | - | 10 - 11 | 795 | 131 | |
BC035090.2 | - | 10 - 11 | 1187 | 131 | |
D89618.1 | - | 10 - 11 | 862 | 131 |