Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KPNA3
Entrez Gene ID
3839
Gene Name
karyopherin subunit alpha 3
Gene Aliases
IPOA4, SRP1, SRP1gamma, SRP4, hSRP1
Chromosome Location
Chr.13: 49699307 - 49792921 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002267.3 NP_002258.2 15 - 16 1783 159
XM_017020561.1 XP_016876050.1 15 - 16 1466 159
GenBank mRNA AF005263.1 - 15 - 16 1380 159
AF034756.1 - 15 - 16 1370 159
AK223480.1 - 15 - 16 1405 159
AK290528.1 - 15 - 16 1443 159
AK291000.1 - 15 - 16 1407 159
BC017355.2 - 15 - 16 1392 159
BC024202.2 - 15 - 16 1392 159
BC035090.2 - 15 - 16 1784 159
D89618.1 - 15 - 16 1459 159
Y12394.1 - 15 - 16 1383 159

View Full Product Details